missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluenesulfonic Acid, 12% in Acetic Acid, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 421222500

 View more versions of this product

Catalog No. AC421222500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Toluenesulfonic acid
12% in Acetic Acid
116.0°C
MFCD00064387
11 to 13% (Alkalimetry)
1.0700g/mL
p-toluenesulfonic acid, p-toluenesulphonic acid, 4-toluenesulfonic acid, tosic acid, p-tolylsulfonic acid, toluene-4-sulfonic acid, toluenesulfonic acid, tosylic acid, benzenesulfonic acid, 4-methyl, p-methylbenzenesulfonic acid
JOXIMZWYDAKGHI-UHFFFAOYSA-N
4-methylbenzenesulfonic acid
6101
172.2
Glass bottle
1.07
41°C
Colorless to Yellow
104-15-4,64-19-7
C7H8O3S
11, 97
01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
15, 9692
Solubility in water: soluble.
CC1=CC=C(C=C1)S(=O)(=O)O
172.198
CHEBI:27849
Pure
1.3825 to 1.3845
250 g
Solution
Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only