missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 171785000

 View more versions of this product

Catalog No. AC171785000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Toluenesulfonic acid monohydrate
100.0°C to 108.0°C
180°C
97.5%
C7H8O3S·H2O
MFCD00142137
11, 97
15, 9692
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and chloroform
CC1=CC=C(C=C1)S(=O)(=O)O.O
190.22
190.22
Pure
6192-52-5
Pink to White
Authentic
Plastic bottle
CH3C6H4SO3H·H2O
500 g
01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
KJIFKLIQANRMOU-UHFFFAOYSA-N
4-methylbenzenesulfonic acid;hydrate
521998
97.5%
Agglomerating Crystals

Chemical Identifiers

6192-52-5
KJIFKLIQANRMOU-UHFFFAOYSA-N
521998
CC1=CC=C(C=C1)S(=O)(=O)O.O
MFCD00142137
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
4-methylbenzenesulfonic acid;hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

Recommended Storage : Product becomes pink on ageing

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only