missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Toluenesulfonic acid sodium salt, 90+%, Thermo Scientific Chemicals
$58.74 - $159.62
Chemical Identifiers
CAS | 657-84-1 |
---|---|
Molecular Formula | C7H7NaO3S |
Molecular Weight (g/mol) | 194.18 |
MDL Number | MFCD00798566,MFCD00064388 |
InChI Key | KVCGISUBCHHTDD-UHFFFAOYSA-M |
Synonym | sodium p-toluenesulfonate, sodium 4-methylbenzenesulfonate, sodium tosylate, p-toluenesulfonic acid sodium salt, sodium toluenesulfonate, sodium toluenesulphonate, tosylate, sodium, naxonate hydrotrope, cyclophil sts 70, eltesol st 34 |
PubChem CID | 3720192 |
IUPAC Name | sodium;4-methylbenzenesulfonate |
SMILES | [Na+].CC1=CC=C(C=C1)S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1933218
|
Thermo Scientific Chemicals
A1933218 |
50 g |
Each for $58.74
|
|
|||||
AAA1933230
|
Thermo Scientific Chemicals
A1933230 |
250 g |
Each for $159.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
657-84-1 | |
194.18 | |
KVCGISUBCHHTDD-UHFFFAOYSA-M | |
3720192 | |
[Na+].CC1=CC=C(C=C1)S([O-])(=O)=O |
C7H7NaO3S | |
MFCD00798566,MFCD00064388 | |
sodium p-toluenesulfonate, sodium 4-methylbenzenesulfonate, sodium tosylate, p-toluenesulfonic acid sodium salt, sodium toluenesulfonate, sodium toluenesulphonate, tosylate, sodium, naxonate hydrotrope, cyclophil sts 70, eltesol st 34 | |
sodium;4-methylbenzenesulfonate |
Specifications
657-84-1 | |
MFCD00798566,MFCD00064388 | |
sodium p-toluenesulfonate, sodium 4-methylbenzenesulfonate, sodium tosylate, p-toluenesulfonic acid sodium salt, sodium toluenesulfonate, sodium toluenesulphonate, tosylate, sodium, naxonate hydrotrope, cyclophil sts 70, eltesol st 34 | |
[Na+].CC1=CC=C(C=C1)S([O-])(=O)=O | |
194.18 | |
194.19 (Anhydrous) | |
50 g |
C7H7NaO3S | |
14,9533 | |
KVCGISUBCHHTDD-UHFFFAOYSA-M | |
sodium;4-methylbenzenesulfonate | |
3720192 | |
≥90% | |
p-Toluenesulfonic acid sodium salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319-H500
TSCA : Yes
RUO – Research Use Only