missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Toluenesulfonyl Chloride, 99%, Spectrum™ Chemical
$178.65 - $178.65
Chemical Identifiers
CAS | 98-59-9 |
---|---|
Molecular Formula | C7H7ClO2S |
Molecular Weight (g/mol) | 190.64 |
MDL Number | MFCD00007450 |
InChI Key | YYROPELSRYBVMQ-UHFFFAOYSA-N |
IUPAC Name | 4-methylbenzene-1-sulfonyl chloride |
SMILES | CC1=CC=C(C=C1)S(Cl)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18612983
|
Spectrum Chemical Mfg Cor
T1094100GM |
100 g | Amber Glass Bottle |
Each for $178.65
|
|
||||
Chemical Identifiers
98-59-9 | |
190.64 | |
YYROPELSRYBVMQ-UHFFFAOYSA-N | |
CC1=CC=C(C=C1)S(Cl)(=O)=O |
C7H7ClO2S | |
MFCD00007450 | |
4-methylbenzene-1-sulfonyl chloride |
Specifications
67° to 69°C | |
1 | |
MFCD00007450 | |
CC1=CC=C(C=C1)S(Cl)(=O)=O | |
190.64 | |
Reagent | |
Amber Glass Bottle |
98-59-9 | |
C7H7ClO2S | |
YYROPELSRYBVMQ-UHFFFAOYSA-N | |
4-methylbenzene-1-sulfonyl chloride | |
99% | |
100 g |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.