missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluic hydrazide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1260414

 View more versions of this product

Catalog No. AAA1260414


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Toluic hydrazide
115°C to 118°C
MFCD00007607
607602
MFFVZXOPRXMVET-UHFFFAOYSA-N
4-methylbenzohydrazide
77174
98%
3619-22-5
C8H10N2O
25 g
p-toluic hydrazide, p-toluic acid hydrazide, p-toluoylhydrazine, 4-methylbenzoylhydrazine, p-toluic acid, hydrazide, 4-methylbenzoic acid hydrazide, p-methylbenzohydrazide, p-toluohydrazide, p-methylbenzoylhydrazine, 4-toluic acid hydrazide
CC1=CC=C(C=C1)C(=O)NN
150.18
150.18

Chemical Identifiers

3619-22-5
150.18
MFFVZXOPRXMVET-UHFFFAOYSA-N
77174
CC1=CC=C(C=C1)C(=O)NN
C8H10N2O
MFCD00007607
p-toluic hydrazide, p-toluic acid hydrazide, p-toluoylhydrazine, 4-methylbenzoylhydrazine, p-toluic acid, hydrazide, 4-methylbenzoic acid hydrazide, p-methylbenzohydrazide, p-toluohydrazide, p-methylbenzoylhydrazine, 4-toluic acid hydrazide
4-methylbenzohydrazide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 222-815-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only