missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 376350010

 View more versions of this product

Catalog No. AC376350010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Toluidine
106-49-0
White to Brown
87°C
98.5% min. (GC)
99%
CH3C6H4NH2
15,9696
Solubility in water: 1.1 g/100mL. Other solubilities: freely soluble in diluted acids, freely soluble in alcohol, ether, acetone, carbon, disulfide, methanol and oils, 156% in ethanol
CC1=CC=C(C=C1)N
107.16
CHEBI:37825
99%
99%
41.0°C to 46.0°C
200.0°C
Authentic
Glass bottle
C7H9N
1 kg
p-toluidine, 4-toluidine, p-tolylamine, 4-aminotoluene, benzenamine, 4-methyl, 4-methylbenzenamine, p-methylaniline, p-methylbenzenamine, p-toluidin, p-aminotoluene
RZXMPPFPUUCRFN-UHFFFAOYSA-N
4-methylaniline
7813
107.16
Crystalline Mass or Crystals

Chemical Identifiers

106-49-0
107.16
p-toluidine, 4-toluidine, p-tolylamine, 4-aminotoluene, benzenamine, 4-methyl, 4-methylbenzenamine, p-methylaniline, p-methylbenzenamine, p-toluidin, p-aminotoluene
CHEBI:37825
CC1=CC=C(C=C1)N
C7H9N
RZXMPPFPUUCRFN-UHFFFAOYSA-N
7813
4-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
Toxic if inhaled.
Very toxic to aquatic life.
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 203-403-1

Recommended Storage : Product may darken on storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only