missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluoyl chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 147980050

 View more versions of this product

Catalog No. AC147980050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Toluoyl chloride
874-60-2
Colorless to Brown
225.0°C to 227.0°C
Authentic
Glass bottle
1.5525 to 1.5545
MFCD00000696
09,486
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
NQUVCRCCRXRJCK-UHFFFAOYSA-N
4-methylbenzoyl chloride
13405
98%
98%
-2.0°C
1.1600g/mL
82°C
98%
C8H7ClO
CH3C6H4COCl
5 mL
1.16
Solubility in water: decomposes.
CC1=CC=C(C=C1)C(=O)Cl
154.6
154.6
Liquid

Chemical Identifiers

874-60-2
154.6
NQUVCRCCRXRJCK-UHFFFAOYSA-N
13405
CC1=CC=C(C=C1)C(=O)Cl
C8H7ClO
MFCD00000696
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
4-methylbenzoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 212-864-8

RTECSNumber : XV1660000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only