missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Tolylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 139160250

 View more versions of this product

Catalog No. AC139160250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Tolylhydrazine hydrochloride
241°C
Authentic
Glass bottle
CH3C6H4NHNH2·HCl
25 g
p-tolylhydrazine hydrochloride, 4-methylphenylhydrazine hydrochloride, 4-methylphenyl hydrazine hydrochloride, 4-tolylhydrazine hydrochloride, hydrazine, 4-methylphenyl-, monohydrochloride, p-tolylhydrazinium 1+ chloride, hydrazine, p-tolyl-, hydrochloride, 4-methylphenylhydrazine hcl, 1-4-methylphenyl hydrazine hydrochloride, p-tolyl-hydrazine hydrochloride
HMHWNJGOHUYVMD-UHFFFAOYSA-N
(4-methylphenyl)hydrazine;hydrochloride
12504
98%
637-60-5
Beige-Brown to White
98%
C7H10N2·HCl
MFCD00012940
15, 510
Solubility in water: soluble
CC1=CC=C(C=C1)NN.Cl
158.63
158.63
Flaky Powder

Chemical Identifiers

637-60-5
158.63
HMHWNJGOHUYVMD-UHFFFAOYSA-N
12504
CC1=CC=C(C=C1)NN.Cl
C7H10N2·HCl
MFCD00012940
p-tolylhydrazine hydrochloride, 4-methylphenylhydrazine hydrochloride, 4-methylphenyl hydrazine hydrochloride, 4-tolylhydrazine hydrochloride, hydrazine, 4-methylphenyl-, monohydrochloride, p-tolylhydrazinium 1+ chloride, hydrazine, p-tolyl-, hydrochloride, 4-methylphenylhydrazine hcl, 1-4-methylphenyl hydrazine hydrochloride, p-tolyl-hydrazine hydrochloride
(4-methylphenyl)hydrazine;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-295-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only