missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal, Acros Organics, Quantity: 800mL, Packaging: AcroSeal Glass bottle, Molecular Weight: 195.34, CAS: 4294-57-9, 60-29-7, Density: 0.8660g/mL, Flash Point: -40 deg.C, Formula Weight: 195.34

$124.00 - $124.00

Chemical Identifiers

CAS 4294-57-9
Molecular Formula C7H7BrMg
Molecular Weight (g/mol) 195.34
MDL Number MFCD00000040
InChI Key ZRJNGFJIBZKXTP-UHFFFAOYSA-M
Synonym p-tolylmagnesium bromide, grignard reagent, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-tolyl magnesium bromide, p-methylphenylmagnesium bromide, p-methylphenylmagnesium bromide
PubChem CID 3644322
IUPAC Name magnesium;methylbenzene;bromide
SMILES CC1=CC=C([Mg]Br)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC426761000
SDS View Documents
Thermo Scientific Chemicals
426761000
100 mL AcroSeal™ Glass bottle
Each for $124.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4294-57-9
195.34
ZRJNGFJIBZKXTP-UHFFFAOYSA-M
3644322
CC1=CC=C([Mg]Br)C=C1
C7H7BrMg
MFCD00000040
p-tolylmagnesium bromide, grignard reagent, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-tolyl magnesium bromide, p-methylphenylmagnesium bromide, p-methylphenylmagnesium bromide
magnesium;methylbenzene;bromide

Specifications

4294-57-9,60-29-7
-40°C
C7H7BrMg
MFCD00000040
0.866
ZRJNGFJIBZKXTP-UHFFFAOYSA-M
magnesium;methylbenzene;bromide
3644322
p-Tolylmagnesium bromide
0.8660g/mL
AcroSeal™ Glass bottle
CH3C6H4MgBr
100 mL
p-tolylmagnesium bromide, grignard reagent, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-tolyl magnesium bromide, p-methylphenylmagnesium bromide, p-methylphenylmagnesium bromide
CC1=CC=C([Mg]Br)C=C1
195.34
195.34
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only