missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Xylene-2-sulfonic Acid Hydrate 98.0+%, TCI America™
Supplier: TCI America D0681100G
Specifications
p-Xylene-2-sulfonic Acid Hydrate | |
609-54-1 | |
100 g | |
C8H10O3S | |
p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641 | |
CC1=CC=C(C)C(=C1)S(O)(=O)=O | |
186.23 | |
186.23 |
86°C | |
Red-White | |
Crystal-Powder at 20°C | |
MFCD00151010 | |
IRLYGRLEBKCYPY-UHFFFAOYSA-N | |
2,5-dimethylbenzene-1-sulfonic acid | |
11868 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
609-54-1 | |
186.23 | |
IRLYGRLEBKCYPY-UHFFFAOYSA-N | |
11868 | |
CC1=CC=C(C)C(=C1)S(O)(=O)=O |
C8H10O3S | |
MFCD00151010 | |
p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641 | |
2,5-dimethylbenzene-1-sulfonic acid |
Safety and Handling
EINECSNumber : (9)-2093
TSCA : Yes