missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Xylene, 99+%, extra pure, Thermo Scientific Chemicals

$186.10 - $433.26

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC220210010
SDS View Documents
Thermo Scientific Chemicals
220210010
1 L Glass Bottle
Each for $186.10
 
AC220210025
SDS View Documents
Thermo Scientific Chemicals
220210025
2.5 L Glass Bottle
Each for $433.26
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-42-3
106.17
URLKBWYHVLBVBO-UHFFFAOYSA-N
7809
1,4-xylene
C8H10
MFCD00008556
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
CHEBI:27417
CC1=CC=C(C)C=C1

Specifications

13.0°C
0.8660g/mL
1 L
Authentic
Liquid
0.005% max.
0.866
C6H4(CH3)2
0.648 mPa.s (20°C)
0.05% max. (Coulometric)
15, 10278
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
CC1=CC=C(C)C=C1
106.17
CHEBI:27417
≥99%
p-Xylene, Extra Pure
106-42-3
138.0°C
25°C
Glass Bottle
1.4940 to 1.4960
99+%
C8H10
MFCD00008556
1307
05, 382
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
URLKBWYHVLBVBO-UHFFFAOYSA-N
1,4-xylene
7809
106.17
Extra Pure
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only