missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Xylene, 99%, for HPLC, Thermo Scientific Chemicals

$45.77 - $319.46

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC445681000
SDS View Documents
Thermo Scientific Chemicals
445681000
100 mL Glass Bottle
Each for $45.77
 
AC445680010
SDS View Documents
Thermo Scientific Chemicals
445680010
1 L Glass Bottle
Each for $223.07
 
AC445680025
SDS View Documents
Thermo Scientific Chemicals
445680025
2.5 L Glass Bottle
Each for $319.46
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-42-3
106.17
URLKBWYHVLBVBO-UHFFFAOYSA-N
7809
1,4-xylene
C8H10
MFCD00008556
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
CHEBI:27417
CC1=CC=C(C)C=C1

Specifications

13.0°C
0.01 max. at 380nm, 0.01 max. at 400nm, 0.05 max. at 350nm, 0.1 max. at 320nm, 1 max. at 294nm
138.0°C
25°C
1.4940 to 1.4960
0.866
C6H4(CH3)2
0.648 mPa.s (20°C)
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
CC1=CC=C(C)C=C1
106.17
CHEBI:27417
99%
p-Xylene
106-42-3
0.8660g/mL
100 mL
Glass Bottle
99%
C8H10
MFCD00008556
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
URLKBWYHVLBVBO-UHFFFAOYSA-N
1,4-xylene
7809
106.17
HPLC
Videos
Safety and Handling

Safety and Handling

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only