Learn More
p-Xylene, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158775000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Xylene | |
106-42-3 | |
0.8660g/mL | |
500 mL | |
Authentic | |
Liquid | |
99% | |
C8H10 | |
MFCD00008556 | |
05, 382 | |
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
1,4-xylene | |
7809 | |
106.17 | |
Pure |
13.0°C | |
Colorless to Yellow | |
138.0°C | |
25°C | |
Glass Bottle | |
1.4940 to 1.4960 | |
0.866 | |
C6H4(CH3)2 | |
0.648 mPa.s (20°C) | |
15, 10278 | |
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents | |
CC1=CC=C(C)C=C1 | |
106.17 | |
CHEBI:27417 | |
99% |
Chemical Identifiers
106-42-3 | |
106.17 | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
7809 | |
1,4-xylene |
C8H10 | |
MFCD00008556 | |
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene | |
CHEBI:27417 | |
CC1=CC=C(C)C=C1 |
Safety and Handling
GHS P Statement
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only