missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Xylene, EMD-Millipore
Supplier: Technidata 1097990005
Specifications
p-Xylene | |
Glass | |
1.1 to 7 %(V) | |
8.7 hPa (20°C) | |
Density: 0.86g/cm3 (20°) | |
C6H4(CH3)2 | |
1, 4-Dimethylbenzene | |
CC1=CC=C(C)C=C1 | |
106.17 | |
CHEBI:27417 |
106-42-3 | |
5 mL | |
540°C | |
≥99.5% | |
C8H10 | |
MFCD00008556 | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
1,4-xylene | |
7809 | |
106.17 |
Chemical Identifiers
106-42-3 | |
106.17 | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
7809 | |
1,4-xylene |
C8H10 | |
MFCD00008556 | |
1, 4-Dimethylbenzene | |
CHEBI:27417 | |
CC1=CC=C(C)C=C1 |
Safety and Handling
Recommended Storage : Store without restrictions