missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Xylene, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S3840

Catalog No. S3840


Explore available promotions

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
p-Xylene106-42-31000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

p-Xylene
2mL amber prescored glass ampule
0.1,99.9
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
C8H10
URLKBWYHVLBVBO-UHFFFAOYSA-N
1,4-xylene
7809
Methanol-P&T
106-42-3,67-56-1
1 mL
0.1,99.9
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
MFCD00008556
CC1=CC=C(C)C=C1
106.17
CHEBI:27417

Chemical Identifiers

106-42-3
106.17
URLKBWYHVLBVBO-UHFFFAOYSA-N
CHEBI:27417
CC1=CC=C(C)C=C1
C8H10
MFCD00008556
7809
1,4-xylene
Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only