Dentec Safety™ P100 Low Profile Cartridge
Dentec Safety™ P100 Low Profile Cartridge is uniquely designed with a space plenum between the filtering material and the inhalation port
Supplier: Dentec Safety™ DTC6451830TLPO
Description
- Inhaled air is evenly distributed across the filter, lowering breathing resistance and extending cartridge life.
- Ultra thin design fits under any grinding shield or welding helmet.
- Will not obstruct field of vision
- Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
- Offset connector improves visibility and allows face shields or welding helmets to be worn without interference
Specifications
Purple | |
Respirator Accessories | |
Cartridges and filters |
Plastic | |
Masks | |
6/box |