Learn More
Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents
Fisherbrand™ Pacific Hemostasis Activated Partial Thromboplastin Time (aPTT) Reagents are manufactured with up to two years extended shelf life.
$145.97 - $234.53
Numéro de catalogue | Numéro du manufacturier. | Description | Quantity | Prix | Quantité | ||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Description | Quantity | Prix | Quantité | ||||
CMS-285-545
|
Thermo Scientific™
100304 |
CaCl2, 0.02M | 10 x 10 mL |
chaque for $145.97
|
|
||||
CMS022929
|
Thermo Scientific™
100402 |
APTT-XL (Ellagic Acid Activator) | 10 x 4 mL |
paquet de 10 chaque for $167.38
|
|
||||
CMS-022-927
|
Thermo Scientific™
100403 |
aPTT-XL | 10 x 10 mL |
chaque for $234.53
|
|
||||
Description
Identify acquired or congenital disorders to the intrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Heparin, reagents are manufactured with up to two years extended shelf life and have excellent lot-to-lot consistency.
Features:- 10mL vials can complete â˽1000 full volume, single determinations; 4mL vials can complete â˽400 full volume, single determinations
- Open vial stability: 30 days at 2° to 8°C
aPTT-XL (Ellagic Acid Activator) (Product No. 10-0402):
- Superior factor sensitivity and responsiveness
- Increased sensitivity to heparin and lupus-like inhibitors
KONTACT™ (Particulate Activator) (Product No. 01-0402):
- Higher sensitivity to Factor VIII
- Average Normal Values: 26 - 33 Seconds
Spécifications
Activated Partial Thromboplastin Time (APTT) |