Thermo Scientific™ Pacific Hemostasis™ Imidazole Buffered Saline
Fisherbrand™ Pacific Hemostasis Fibrinogen/Fibrin Analysis is a complete line of assays used to screen and quantify plasma fibrinogen based on the Clauss method.
Supplier:  Thermo Scientific™ 100600
 View more versions of this product
Catalog No. 22-176-271
Explore available promotions
Description
Description
Screen and quantify plasma fibrinogen based on the Clauss method with Thermo Scientific™ Pacific Hemostasis™ Fibrinogen/Fibrin Analysis.
Fibrinogen Assay Set:
  • Quantifies plasma fibrinogen levels
  • Detects acquired and congenital hemorrhagic disorders
  • Components available separately
Thrombin Time Assay:
  • Provides qualitative determination
  • Useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot
  • Reconstituted stability: 8 hours at 2° to 8°C
Ordering Information:
Products can be purchased as a set or individually.
Specifications
Specifications
2°C to 8°C
Fibrinogen Assay Set
Fibrinogen Assay
100 Determinations