missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ PageRuler™ Plus Prestained Protein Ladder, 10 to 250 kDa

Thermo Scientific™ PageRuler Plus Prestained Protein Ladder is a mixture of nine (9) blue-, orange- and green-stained proteins (10 to 250 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.

Supplier:  Thermo Scientific™ 26619

 View more versions of this product

Catalog No. PI26619

Explore available promotions

Description

Description

Thermo Scientific PageRuler Plus Prestained Protein Ladder is a mixture of nine blue-, orange-, and green-stained proteins (10 to 250 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on SDS-polyacrylamide gels and western blots
Specifications

Specifications

Contents: two vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

9
Yes
3 colors: Blue, Orange, Green
250, 130, 100, 70, 55, 35, 25, 15, 10 kDa
2 x 250 μL
Western Blotting, SDS-PAGE
Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm)
Protein Ladder
10 to 250 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
PageRuler™
Approved for shipment on Wet or Dry Ice
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.