Learn More
Thermo Scientific™ PageRuler™ Prestained Protein Ladder, 10 to 180 kDa
Thermo Scientific PageRuler Prestained Protein Ladder is a mixture of 10 blue-, orange-, and green-stained proteins (10 to 180 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting.
$350.75 - $1,115.49
Specifications
Detection Method | Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm) |
---|---|
Number of Markers | 10 |
Product Type | Protein Ladder |
Ready to Load | Yes |
Size Range | 10 to 180 kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
PI26616
|
Thermo Scientific™
26616 |
2 x 250 μL |
N/A
|
|
||||||
PI26617
|
Thermo Scientific™
26617 |
10 x 250 μL |
N/A
|
|
||||||
Description
Thermo Scientific PageRuler Prestained Protein Ladder is a mixture of 10 blue-, orange-, and green-stained proteins (10 to 180 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Applications
- Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
- Monitoring protein transfer onto membranes after western blotting
- Sizing of proteins on SDS-polyacrylamide gels and western blots
Specifications
Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm) | |
Protein Ladder | |
10 to 180 kDa | |
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels | |
PageRuler™ |
10 | |
Yes | |
3 colors: Blue, Orange, Green | |
180, 130, 100, 70, 55, 40, 35, 25, 15, 10 kDa | |
Approved for shipment on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.