missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ PageRuler™ Unstained Low Range Protein Ladder

Thermo Scientific PageRuler Unstained Low Range Protein Ladder is a mixture of eight purified proteins and peptides (3.4 to 100 kDa) for use as size standards in peptide and small-protein gel electrophoresis.

Supplier:  Thermo Scientific™ 26632

 View more versions of this product

Catalog No. PI26632


Explore available promotions

Description

Description

Thermo Scientific™ PageRuler Unstained Low Range Protein Ladder is a mixture of eight (8) purified proteins and peptides (3.4 to 100kDa) for use as size standards in peptide and small protein gel electrophoresis.

Thermo Scientific PageRuler Unstained Low Range Protein Ladder is a mixture of eight purified proteins and peptides (3.4 to 100 kDa) for use as size standards in peptide and small-protein gel electrophoresis. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Product features

  • Reference bands—25 kDa band is stronger than the others for easy orientation
  • Tagged—the ladder proteins (except for the 5 and 3.4 kDa peptides) contains an integral Strep-tag™ II sequence and can be detected on western blots using Strep-Tactin™ conjugates

Applications

  • Accurate protein sizing on SDS-polyacrylamide gels and western blots
Specifications

Specifications

Includes two vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 100 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol

Upon receipt, store at -20°C.
Protein Ladder
Tricine gels
Unstained
100, 30, 25, 20, 15, 10, 5, 3.4 kDa
2 x 250 μL
SDS-PAGE
8
Yes
3.4 to 100 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
PageRuler™
Approved for shipment on Wet or Dry Ice
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.