missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Painted Steel Mobile Cabinet
Steel mobile cabinets are on locking swivel casters which allow them to be moved. Ideal for under a table or workstation lab environment.
Supplier: Fisherbrand™ FJTP102130P
Description
- Cabinet has a finished back and top
- Provided without locks
- Counter weighted anti-tip design
- Optional work surface sold separately
Specifications
30 in. | |
22 in. | |
Yes | |
76.2 cm | |
Mobile Cabinet | |
White |
3 | |
81.5 cm | |
55.9 cm | |
32.1 in. | |
None | |
Steel |