Learn More
Palladium, 99.99%, (trace metal basis), powder, particle size -20 mesh, Thermo Scientific Chemicals
$938.16 - $12178.10
Chemical Identifiers
CAS | 7440-05-3 |
---|---|
Molecular Formula | Pd |
Molecular Weight (g/mol) | 106.42 |
MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
Synonym | black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon |
PubChem CID | 23938 |
ChEBI | CHEBI:33363 |
IUPAC Name | palladium |
SMILES | [Pd] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC193690010
|
Thermo Scientific Chemicals
193690010 |
1 g | Glass bottle |
Each for $938.16
|
|
||||
AC193690250
|
Thermo Scientific Chemicals
193690250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7440-05-3 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Specifications
Black to Gray | |
7440-05-3 | |
99.99% | |
Pd | |
1 g | |
15, 7090 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
106.4 | |
Trace Metal Basis | |
Powder |
1555.0°C | |
2970.0°C | |
Pd | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
99.99% | |
Glass bottle | |
Palladium |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only