missing translation for 'onlineSavingsMsg'
Learn More

Palladium on activated carbon, 10% Pd, unreduced, dry

CAS: 7440-05-3 | Pd | 106.42 g/mol

$74.02 - $3718.86

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC195030010
SDS View Documents
Thermo Scientific Chemicals
195030010
1 g Glass bottle
Each for $74.02
Only null left
 
AC195030500
SDS View Documents
Thermo Scientific Chemicals
195030500
50 g Glass Bottle
Each for $3,718.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]

Specifications

Black
1 g
Particle size distribution: ≤4 Micron: 10%, ≤21 Micron: 50%, ≤90 Micron: 90%
Pd
01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
10% (Pd)
7440-05-3,7440-44-0
Glass bottle
Powder
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
15, 7090
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
106.42
Palladium on activated carbon, 10% Pd
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only