missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium on alumina, 5% Pd, powder, dry, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 293030500

 View more versions of this product

Catalog No. AC293030500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Black or Gray to Silver
7440-05-3
Pd
50 g
15, 7090
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
106.42
Glass bottle
Palladium on alumina
Pd
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
4.0 to 6.0% (Pd)
Powder

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-115-6

RTECSNumber : RT3480500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only