missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium black, 99.9% (metals basis), Thermo Scientific Chemicals

Use as a catalyst in hydrogenation reactions. Other uses include the manufacture of paints, adhesives, fibers and coatings. Basic particles of Palladium black have the tendency to cluster into macroscopic aggregates.

$604.47 - $604.47

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA0065903
View Documents
Thermo Scientific Chemicals
00065903
1 g
Each for $604.47
 
Description

Description

It is used as a catalyst in hydrogenation reactions. Further it finds use in the manufacture of paints, adhesives, fibers and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]

Specifications

Gray to Silver
7440-05-3
Powder
99.9% (Metals basis)
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Odorless
Palladium black
1555°C
3067°C
12.02g/cm3 at 20°C
Pd
1 g
Insoluble in water; Soluble in aqua regia and fused alkalies
[Pd]
106.42
CHEBI:33363
Powder
Videos
Safety and Handling

Safety and Handling

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-115-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only