missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Form: Approx.0.2g/25 x 25mm

Supplier:  Thermo Scientific Chemicals 011517GH

 View more versions of this product

Catalog No. AA11517GH


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver to Gray
7440-05-3
12.02g/cm3 at 20°C
Pd
100 x 100 mm
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Odorless
Palladium
Silver gray foil
99.9% (Metals basis)
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
Foil

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-115-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only