missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Palladium Silver foil, 0.025mm (0.001in) thick, Thermo Scientific Chemicals
$509.51 - $509.51
Specifications
Chemical Name or Material | Palladium Silver foil |
---|---|
Name Note | 0.025mm (0.001 in.) thick |
Avg. Mol. Wt. or Mol. Wt. Range | Pd:Ag; 77:23 wt% |
Assay Percent Notes | (metals basis excluding Pt) |
MDL Number | MFCD02091738 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA45003FF
|
Thermo Scientific Chemicals
045003FF |
25 x 25 mm |
Each for $509.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Palladium Silver foil | |
Pd:Ag; 77:23 wt% | |
MFCD02091738 | |
Odorless | |
Foil |
0.025mm (0.001 in.) thick | |
(metals basis excluding Pt) | |
99.9% | |
≈0.18g/25x25mm |
RUO – Research Use Only