missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Palladium wire, 0.25mm (0.01in) dia, 99.97% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 043738BV
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Palladium wire | |
7440-05-3 | |
Pd | |
30 cm | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
99.97% | |
≈0.6g/m |
0.25mm (0.01 in.) dia. | |
(metals basis) | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
Odorless | |
Wire |
Chemical Identifiers
7440-05-3 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Safety and Handling
EINECSNumber : 231-115-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only