missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Palmitic acid, 98%

Supplier:  Thermo Scientific Chemicals 129702500

 View more versions of this product

Catalog No. AC129702500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Palmitic acid
215 to 233mg KOH/g
White
351.5°C
Authentic
Plastic bottle
CH3(CH2)14CO2H
250 g
02,IV,1157
palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
IPCSVZSSVZVIGE-UHFFFAOYSA-N
hexadecanoic acid
985
CHEBI:15756
98%
57-10-3
59.0°C to 63.0°C
0.8500g/mL
206°C
97.5% min. (GC)
C16H32O2
MFCD00002747
216 to 224mg KOH/g
15,7097
Solubility in water: insoluble. Other solubilities: soluble in hot alcohol, ether and propyl alcohol
CCCCCCCCCCCCCCCC(O)=O
256.43
7.8 mPa.s (70°C)
256.42
Flakes

Chemical Identifiers

57-10-3
256.43
IPCSVZSSVZVIGE-UHFFFAOYSA-N
985
hexadecanoic acid
C16H32O2
MFCD00002747
palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
CHEBI:15756
CCCCCCCCCCCCCCCC(O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Fatal if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-312-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only