missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Palmitic acid, sodium salt, 98%, Thermo Scientific Chemicals
$134.19 - $411.38
Chemical Identifiers
CAS | 408-35-5 |
---|---|
Molecular Formula | C16H31NaO2 |
Molecular Weight (g/mol) | 278.4 |
MDL Number | MFCD00002749 |
InChI Key | GGXKEBACDBNFAF-UHFFFAOYSA-M |
Synonym | sodium palmitate, palmitic acid sodium salt, hexadecanoic acid, sodium salt, sodium hexadecanoate, palmitic acid, sodium salt, sodium pentadecanecarboxylate, unii-jq43kp6296, hsdb 759, hexadecanoic acid sodium salt, hexadecanoic acid,sodium salt 1:1 |
PubChem CID | 2735111 |
IUPAC Name | sodium;hexadecanoate |
SMILES | CCCCCCCCCCCCCCCC(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC416700050
|
Thermo Scientific Chemicals
416700050 |
5 g | Glass bottle |
Each for $134.19
|
|
||||
AC416700250
|
Thermo Scientific Chemicals
416700250 |
25 g | Glass bottle |
Each for $411.38
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
408-35-5 | |
278.4 | |
GGXKEBACDBNFAF-UHFFFAOYSA-M | |
2735111 | |
CCCCCCCCCCCCCCCC(=O)[O-].[Na+] |
C16H31NaO2 | |
MFCD00002749 | |
sodium palmitate, palmitic acid sodium salt, hexadecanoic acid, sodium salt, sodium hexadecanoate, palmitic acid, sodium salt, sodium pentadecanecarboxylate, unii-jq43kp6296, hsdb 759, hexadecanoic acid sodium salt, hexadecanoic acid,sodium salt 1:1 | |
sodium;hexadecanoate |
Specifications
408-35-5 | |
2% max. (110°C, 4 hrs) | |
98% | |
C16H31NaO2 | |
MFCD00002749 | |
02,III,94 | |
GGXKEBACDBNFAF-UHFFFAOYSA-M | |
sodium;hexadecanoate | |
2735111 | |
98% | |
Palmitic acid, sodium salt |
White | |
Authentic | |
Glass bottle | |
CH3(CH2)14COONa | |
5 g | |
sodium palmitate, palmitic acid sodium salt, hexadecanoic acid, sodium salt, sodium hexadecanoate, palmitic acid, sodium salt, sodium pentadecanecarboxylate, unii-jq43kp6296, hsdb 759, hexadecanoic acid sodium salt, hexadecanoic acid,sodium salt 1:1 | |
CCCCCCCCCCCCCCCC(=O)[O-].[Na+] | |
278.4 | |
278.4 | |
Powder |
Safety and Handling
EINECSNumber : 206-988-1
RUO – Research Use Only