missing translation for 'onlineSavingsMsg'
Learn More

Pamabrom 98.0+%, TCI America™

$29.83 - $59.65

Chemical Identifiers

CAS 606-04-2
Molecular Formula C11H18BrN5O3
Molecular Weight (g/mol) 348.201
MDL Number MFCD00867328
InChI Key ATOTUUBRFJHZQG-UHFFFAOYSA-N
Synonym 8-Bromotheophylline 2-Amino-2-methyl-1-propanol
PubChem CID 11806
IUPAC Name 2-amino-2-methylpropan-1-ol;8-bromo-1,3-dimethyl-7H-purine-2,6-dione
SMILES CC(C)(CO)N.CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P23591G
View Documents
TCI America
P23591G
1 g
Each for $29.83
Only null left
 
P23595G
View Documents
TCI America
P23595G
5 g
Each for $59.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

606-04-2
348.201
ATOTUUBRFJHZQG-UHFFFAOYSA-N
11806
CC(C)(CO)N.CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Br
C11H18BrN5O3
MFCD00867328
8-Bromotheophylline 2-Amino-2-methyl-1-propanol
2-amino-2-methylpropan-1-ol;8-bromo-1,3-dimethyl-7H-purine-2,6-dione

Specifications

606-04-2
White
MFCD00867328
8-Bromotheophylline 2-Amino-2-methyl-1-propanol
CC(C)(CO)N.CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Br
348.201
348.20
Crystalline Powder
295°C
C11H18BrN5O3
1 g
ATOTUUBRFJHZQG-UHFFFAOYSA-N
2-amino-2-methylpropan-1-ol;8-bromo-1,3-dimethyl-7H-purine-2,6-dione
11806
≥98.0% (HPLC,T)
Pamabrom
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'