Papain, Powder, Purified, Spectrum™ Chemical
PA125, 9001-73-4
$413.46 - $1193.31
Description
Spectrum™ Chemical Papain, Powder, Purified , is a cysteine protease enzyme the is present in the papaya fruit. It is traditionnally used to break down meat fibers. It has more recently been used to dissociate cells in the early steps of cell culture preparation.Chemical Identifiers
9001-73-4 |
Specifications
Purified | |
Poly Bottle | |
9001-73-4 | |
100 g |