missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cardinal Health™ Paper Sterilization Tubing
For long or odd-shaped instruments
Supplier: Cardinal Health™ T90003
Description
- Fracture-resistant film
- High-strength
- Surgical-grade
- Provides adequate breathability and blue required sterile barrier
- Peelable, low-lint design
- Dual chemical indicators identify steam or EO sterilization
- Length: 100 ft. (30.5m)
Specifications
Low-lint | |
Long or odd shaped instrument | |
10/Cs. | |
100 ft. | |
3 in. |
Blue | |
Sterile | |
100 ft. x 3 in. (30.5m x 7.6 cm) | |
30.5 m | |
7.6 cm |