Learn More
Paraffin, liquid, pure, Thermo Scientific Chemicals
$62.71 - $245.20
Chemical Identifiers
CAS | 8012-95-1 |
---|---|
Molecular Formula | MFCD00131611 |
Molecular Weight (g/mol) | 0.00 |
MDL Number | MFCD00131611 |
InChI Key | FFNDMZIBVDSQFI-UHFFFAOYSA-N |
Synonym | delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride |
PubChem CID | 68245 |
ChEBI | CHEBI:38701 |
IUPAC Name | 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride |
SMILES | * |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC171402500
|
Thermo Scientific Chemicals
171402500 |
250 mL | Glass bottle |
Each for $62.71
|
|
||||
AC171400010
|
Thermo Scientific Chemicals
171400010 |
1 L | Glass bottle |
Each for $133.51
|
|
||||
AC171400025
|
Thermo Scientific Chemicals
171400025 |
2.5 L | Glass bottle |
Each for $245.20
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
8012-95-1 | |
0.00 | |
FFNDMZIBVDSQFI-UHFFFAOYSA-N | |
68245 | |
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride |
MFCD00131611 | |
MFCD00131611 | |
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride | |
CHEBI:38701 | |
* |
Specifications
8012-95-1 | |
Colorless | |
300.0°C | |
Authentic | |
MFCD00131611 | |
250 mL | |
14, 7186 | |
Solubility in water: insoluble | |
* | |
0.00 | |
65-75 mm2/s (40°C) | |
Pure | |
Paraffin, liquid, Pure |
-24.0°C | |
0.8800g/mL | |
218°C | |
Glass bottle | |
MFCD00131611 | |
0.88 | |
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride | |
FFNDMZIBVDSQFI-UHFFFAOYSA-N | |
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride | |
68245 | |
CHEBI:38701 | |
Oily Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 232-384-2
RUO – Research Use Only