Mopec™ Paraffin Wax Dispenser
The Paraffin Wax Dispenser is designed to take up minimal bench-top space while providing a rapid heating system that produces melted paraffin wax in only 30 minutes.
$10250.94 - $17568.74
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
22-445-493
|
Mopec™
BK575 |
7.5 L |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
22-445-660
|
Mopec™
BK575X2 |
15 L |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Designed to take up minimal bench-top space
- Digital temperature control
- 15 L dispenser has 2 separate tanks with independent heating controls
- Rapid heating system ensures system is ready to use in less time
- Heated tap prevents wax buildup
- Non-drip lever with three positions
- Double-layered tanks provide internal temperature control and ensures outer surfaces are safe to touch
- Anti-microbial coating to help prevent bacteria and virus growth
Specifications
Paraffin Wax Dispenser |