missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paraffin, pure, wax, granular
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure | CAS: 8002-74-2 | CnH2n+2 | 341.451 g/mol
$180.39 - $745.89
Chemical Identifiers
| CAS | 8002-74-2 |
|---|---|
| Molecular Formula | CnH2n+2 |
| Molecular Weight (g/mol) | 341.451 |
| MDL Number | MFCD00132833 |
| InChI Key | JWHAUXFOSRPERK-UHFFFAOYSA-N |
| Synonym | propafenone, rythmol, propafenona, propafenonum, propafenonum inn-latin, propafenona inn-spanish, propafenone inn:ban, gnf-pf-4594, propafenone-hcl, propafenone inn |
| PubChem CID | 4932 |
| ChEBI | CHEBI:63619 |
| IUPAC Name | 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one |
| SMILES | CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC416770020
|
Thermo Scientific Chemicals
416770020 |
2 kg |
Each for $180.39
|
|
|||||
|
AC416770100
|
Thermo Scientific Chemicals
416770100 |
10 kg |
Each for $745.89
|
|
|||||
Description
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure
Chemical Identifiers
| 8002-74-2 | |
| 341.451 | |
| JWHAUXFOSRPERK-UHFFFAOYSA-N | |
| 4932 | |
| 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one |
| CnH2n+2 | |
| MFCD00132833 | |
| propafenone, rythmol, propafenona, propafenonum, propafenonum inn-latin, propafenona inn-spanish, propafenone inn:ban, gnf-pf-4594, propafenone-hcl, propafenone inn | |
| CHEBI:63619 | |
| CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O |
Specifications
| 8002-74-2 | |
| White | |
| 198°C | |
| Plastic bottle | |
| MFCD00132833 | |
| 14,7023 | |
| Solubility in water: insoluble. Other solubilities: insoluble in alcohol (soluble in warm alcohol),insoluble in acids,soluble in benzene,chloroform,ether,carbon,disulfide,ligroin,turpentine,miscible when melted with wax,fats | |
| CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O | |
| 341.451 | |
| 3-4.5 mPa.s (98.9°C) | |
| Pure | |
| Paraffin wax |
| 58.0°C to 62.0°C | |
| 322.0°C | |
| Authentic | |
| CnH2n+2 | |
| 2 kg | |
| propafenone, rythmol, propafenona, propafenonum, propafenonum inn-latin, propafenona inn-spanish, propafenone inn:ban, gnf-pf-4594, propafenone-hcl, propafenone inn | |
| JWHAUXFOSRPERK-UHFFFAOYSA-N | |
| 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one | |
| 4932 | |
| CHEBI:63619 | |
| Pellets |
Safety and Handling
EINECSNumber : 232-315-6
RTECSNumber : RV0350000
TSCA : TSCA
RUO – Research Use Only