missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS

Paraformaldehyde, CH2O, CAS Number-30525-89-4, methylene oxide, paraformaldehyde, formalin, paraform, methyl aldehyde, formic aldehyde, formol, oxymethylene, methanal, oxomethane, 500mL, CHEBI:16842, H317-H318-H351

Supplier:  Thermo Scientific Chemicals J61899AP

 View more versions of this product

Catalog No. AAJ61899AP


Explore available promotions

Description

Description

Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with brass, steel, copper, acid anhydrides, strong oxidizing agents and strong reducing agents.
Specifications

Specifications

Liquid
30525-89-4
CH2O
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
Paraformaldehyde
500 mL
MFCD00133991
Miscible with water.
C=O
30.026
CHEBI:16842

Chemical Identifiers

30525-89-4
30.026
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00133991
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
CHEBI:16842
C=O
Videos
Safety and Handling

Safety and Handling

Causes serious eye damage, Suspected of causing cancer, May cause an allergic skin reaction

Serious eye damage/eye irritation (category 1), Carcinogenicity (category 2), Skin sensitisation (category 1)

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only