missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Pararosaniline chloride, pure, high purity biological stain

Supplier:  Thermo Scientific Chemicals 227885000

 View more versions of this product

Catalog No. AC227885000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pararosaniline chloride
569-61-9
MFCD00001657
Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol
C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl
323.824
CHEBI:87663
Pure
500 g
268.0°C to 270.0°C
C19H18ClN3
Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500
JUQPZRLQQYSMEQ-UHFFFAOYSA-N
4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
11292
323.83
Glass bottle

Chemical Identifiers

569-61-9
323.824
JUQPZRLQQYSMEQ-UHFFFAOYSA-N
11292
4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
C19H18ClN3
MFCD00001657
Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500
CHEBI:87663
C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl
Safety and Handling

Safety and Handling

GHS H Statement:
May cause cancer.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 209-321-2

RTECSNumber : CX9850100

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only