Parker Balston Nitrogen Generators: N2-14 Series from Restek™ DFS Item
Safe, reliable, and requires low maintenance
Supplier:  Chromatographic RK21648
 View more versions of this product
Catalog No. 06711702
Explore available promotions
Description
Description
  • Turns compressed air into ultra-pure nitrogen (up to 99.5%)
  • Model N2-14 requires no electricity
Specifications
Specifications
CE
Prefilters and final filter
Maintenance Kit for Models N2-14, N2-14A, 75-72, 75-720NA
Models N2-14, N2-14A, 75-72, 75-720NA
Turns compressed air into ultra-pure nitrogen