Parr™ 6400 Automatic Isoperibol Calorimeter
High throughput, with minimum operator time between tests
Supplier:  Parr™ 6400CLEA
 View more versions of this product
Catalog No. 04-731-216
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: 6038 maintenance kit, automated bucket and jacket fill, automatic bomb wash, 1138 fixed oxygen bomb of alloy with removable head, FlashROM memory and TCP/IP network communications
Requires : Source of 99.5% oxygen, a source of nitrogen or compressed air at 80psi, and deionized water.
Description
Description
The 6400 incorporates a closed-loop cooling subsystem that consistes of a thermoelectric cooler assembly attached directly to a one liter water tank. An external nitrogen pressurized tank is used to supply rinse water. The 6400 requires one minute of operator time per test, allowing a technician to operate up to four calorimeters simultaneously.
  • Isoperibol calorimetry
  • Automated bucket and jacket fill as well as vessel fill and rinse
  • Automatic cooling
  • 6 to 7 tests per hour
  • Operator time per test is 1 minute
  • 0.1% precision class instrument
  • FlashROM memory and TCP/IP network communications
  • USB port for balance and printer connections
  • Updates via the Internet
1138 Oxygen Bomb
  • Withstands high magnitude testing
  • Head features O-ring groove optimized to minimze frictional wear
  • Removable bomb head allows fast sample loading using proprietary Twist-Lock vessel closure design
Specifications
Specifications
Automatic Isoperibol Calorimeter
Ethernet or USB
40 cm
20 in.
6038 maintenance kit, automated bucket and jacket fill, automatic bomb wash, 1138 fixed oxygen bomb of alloy G30 with removable head, FlashROM memory and TCP/IP network communications, 1576 rinse tank, 1758 printer, one year service kit
1 min./Test
0.001
0.0001°C
115 V
42 cm
ISO 9001:2008, CSA, CE, ASTM
15.5 in.
Measuring heats of combustion of solid or liquid fuels, waste materials and other combustible samples
51 cm
Stores 1000 tests
Automatic
5000 to 8000 calories
6 to 7 Tests/hr.
16.5 in.