Parr™ 6755 Solution Calorimeter
Convenient benchtop, multi-purpose calorimeter
Supplier:  Parr™ 6755EF
 View more versions of this product
Catalog No. 04-731-193
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Utilizing a unique rotating sample cell and a precise microprocessor-based thermometer, the Parr 6755 Solution Calorimeter provides a moderately priced and easily operated instrument for measuring heats of reactions, mixing, solution, dilution and wetting. Measurements are made at ambient temperature and at atmospheric pressure in either liquid-liquid or liquid-solid systems covering energy changes ranging from 2 to 1000 calories.
  • Easy to operate
  • Menu-driven touch screen display
  • Real-time plotting on LCD screen
  • Outputs provided for printer, computer or LAN connection
  • Compensated jacket and Dewar flask bucket
  • Includes 6755 calorimetric thermometer
Specifications
Specifications
Solution Calorimeter
Ethernet or USB
33 cm
Measuring heats of reactions, mixing, solution, dilution and wetting
33 cm
0.004
50°F to 122°F
230 V
22 cm
ISO 9001:2008, CSA, CE, ASTM
13 in.
LCD
13 in.
6772 precision calorimeter thermometer/controller, compensated jacket and dewar flask bucket, SD memory slots and TCP/IP and USB to ethernet LAN network communications
2 to 1000 calories
10°C to 50°C
9 in.