Parr™ Microwave Acid Digestion Bomb
For high-speed heating: 20- to 30-minute cycle time, including cooling.
Supplier:  Parr™ 4782
 View more versions of this product
Catalog No. 04-731-56
Explore available promotions
Alert:
Do not heat in conventional convection oven.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Description
Description
Polymer microwaveable acid digestion bomb for use in any commercial microwave.
  • Operates at temperatures to 250°C (482°F) and pressures to 1200psig (8.3MPa)
  • Use when complete containment of volatile chemicals is required
  • Useful for high-speed heating—20 to 30 minute cycle time, including cooling
  • PTFE cup with cover and self-sealing PTFE O-ring maintains tight seal without heavy preloading
  • Close by hand-turning knurled cap; retaining screw rises farther above top of cap with each 500psig increment of internal pressure
  • No metal parts to corrode
Specifications
Specifications
Microwave Acid Digestion Vessel
45 mL
1200 psi
482°F
1.52 oz.
To dissolve inorganic samples, digest organic materials, other reactions
1 Ea.
250°C