missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Particle-Certified Glass Containers
Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Supplier: Thermo Scientific™ 12108ALP
Description
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Available in clear or amber
- Assembled with low-shedding polypropylene caps with chemically inert PTFE faced liners that do not contain adhesives
- Individual bottles double bagged to maintain integrity until point of use
- 4L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
- Low particle applications
Specifications
8 oz., 250 mL | |
8 oz. | |
Amber Glass Wide Mouth Packer Bottle with Closure, 250 mL | |
Glass | |
Wide | |
Packer Bottle | |
PTFE | |
45-400 mm |
250 mL | |
Amber | |
4.75 in. | |
12 | |
63.5 mm | |
2.5 in. | |
Polypropylene, Low-shedding | |
120.65 mm |