missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Particle-Certified Glass Containers

Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.

Supplier:  Thermo Scientific™ 12216ALP

 View more versions of this product

Catalog No. 12-200-106


Explore available promotions

Description

Description

Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.

Key Features:

  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene caps with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4L jugs suitable for hazardous shipping in conjunction with combination packaging

Recommended for:

  • Low particle applications
Specifications

Specifications

16 oz., 500 mL
16 oz.
Amber Glass Wide Mouth Packer Bottle with Closure, 500 mL
Glass
Wide
Packer Bottle
PTFE
53-400 mm
500 mL
Amber
5.8 in.
12
80.77 mm
3.18 in.
Polypropylene, Low-shedding
147.32 mm
SDS
Documents

Documents

Promotions

Promotions