missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Particle-Certified Glass Containers

Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.

Supplier:  Thermo Scientific™ 11104ALP

 View more versions of this product

Catalog No. 12-200-137


Explore available promotions

Description

Description

Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.

Key Features:

  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene caps with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4L jugs suitable for hazardous shipping in conjunction with combination packaging

Recommended for:

  • Low particle applications
Specifications

Specifications

128 oz., 4000 mL
128 oz.
Amber Glass Jug with Closure, 4000 mL
Glass
336.55 mm
13.25 in.
Polypropylene, Low-shedding
158.75 mm
4000 mL
Amber
6.25 in.
4
Jug
PTFE/Silicone
38-430 mm
SDS
Documents

Documents

Promotions

Promotions