missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Particle-Certified Glass Containers
Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Supplier: Thermo Scientific™ 13216CLP
Description
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Available in clear or amber
- Assembled with low-shedding polypropylene caps with chemically inert PTFE faced liners that do not contain adhesives
- Individual bottles double bagged to maintain integrity until point of use
- 4L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
- Low particle applications
Specifications
16 oz., 500mL | |
16 oz. | |
Clear Glass Wide Mouth Jar with Polypropylene Closure, 500 mL | |
Glass | |
Wide | |
Sample Jar | |
PTFE | |
89-400 mm |
500 mL | |
Clear | |
3.5 in. | |
12 | |
98.4 mm | |
3.875 in. | |
Polypropylene, Low-shedding | |
90 mm |