missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Patent Blue, Spectrum™ Chemical
$262.03 - $1342.12
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18-612-148
|
Spectrum Chemical Mfg Cor
PA13525GM |
25 g | Amber Glass Bottle |
Each for $262.03
|
|
|||||
18-612-147
|
Spectrum Chemical Mfg Cor
PA135100GM |
100 g | Amber Glass Bottle |
Each for $468.68
|
|
|||||
18-612-149
|
Spectrum Chemical Mfg Cor
PA135500GM |
500 g | Amber Glass Bottle |
Each for $1,342.12
|
|
|||||
Chemical Identifiers
129-17-9 |
Specifications
129-17-9 | |
Reagent | |
25 g |
1 | |
Amber Glass Bottle | |
Patent Blue |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.