BD Patient/Exam Room Sharps Collectors
Horizontal, side or vertical entry
$40.23 - $40.23
Specifications
Material Polypropylene
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (English) Color Design Price Quantity  
Catalog Number Mfr. No. Capacity (English) Color Design Price Quantity  
14-826-143
View Documents
BD
300452
3 gal. Yellow Counterbalanced Door
Each for $40.23
Case of 12 Each for $482.76
Description
Description
  • Strong, durable polypropylene construction gives containers excellent impact and puncture-resistance
  • Suitable for incineration or autoclaving
  • Clear top and door provide visual indication of fill level
  • Safety transport handles keep hands away from collector body
Horizontal- and Side-Entry Collectors
  • Counterbalanced door opens automatically for single-handed disposal; stays closed to prevent overfilling
Order Info
Shipped nested; assembly of top to base required.
Specifications
Specifications
Polypropylene
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'