missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega PCR Master Mix DFS Item

Premixed 2X solution of Taq DNA Polymerase, dNTPs and Reaction Buffer.

Supplier:  Promega M7502

 View more versions of this product

Catalog No. PR-M7502


Explore available promotions

Includes: PCR Master Mix, Nuclease-free water

Description

Description

  • Premixed 2X solution of Taq DNA Polymerase, dNTPs and Reaction Buffer
  • One pipetting step adds Taq DNA polymerase, dNTPs, MgCl2 and reaction buffer
  • Set up reactions with as low as 10 μL volume
Specifications

Specifications

• 2X PCR Master Mix, 2 x 1.25 mL
• Nuclease-free Water, 2 x 1.25 mL
• Taq DNA Polymerase
• dNTPs
• MgCl2
• Reaction Buffers

Store at –30° to –10°C.
2X
50 μL
Standard PCR, RT-PCR
100 x 50& mu;L Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions